accessibility menu, dialog, popup

UserName

CAS RN 135-67-1

IUPAC Name: 10H-phenoxazine
Synonyms: phenoxazine
Molecular Weight (g/mol): 183.21
Molecular Formula: C12H9NO
InChi Key: TZMSYXZUNZXBOL-UHFFFAOYSA-N
SMILES: N1C2=CC=CC=C2OC2=CC=CC=C12
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results