missing translation for 'onlineSavingsMsg'
Learn More

Phenoxazine, 97%

Product Code. 10407140 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10407140 5 g Glass bottle 5g
10336870 1 g Glass bottle 1g
10033763 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10407140 Supplier Thermo Scientific Acros Supplier No. 130220050
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 135-67-1 | C12H9NO | 183.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135-67-1
Molecular Formula C12H9NO
Molecular Weight (g/mol) 183.21
MDL Number MFCD00005014
InChI Key TZMSYXZUNZXBOL-UHFFFAOYSA-N
Synonym phenoxazine, phenazoxine, unii-c2zwt499sg, c2zwt499sg, 5,4-oxazine, phenotazine, rarechem fh 2c 0s92, phenoxazine 1g, 9ci
PubChem CID 67278
IUPAC Name 10H-phenoxazine
SMILES N1C2=CC=CC=C2OC2=CC=CC=C12

Specifications

Melting Point 154.0°C to 159.0°C
Color Gray to Gray-Green
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 27, 62
Merck Index 15, 7365
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in abs.methanol, ethanol, ether,, chloroform and benzene, sparingly soluble in petroleum ether
Formula Weight 183.21
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material Phenoxazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.