accessibility menu, dialog, popup

UserName

CAS RN 135050-44-1

IUPAC Name: 3-chloro-4-iodoaniline
Synonyms: 3-chloro-4-iodoaniline
Molecular Weight (g/mol): 253.47
Molecular Formula: C6H5ClIN
InChi Key: ONZHMGRKWJMTDE-UHFFFAOYSA-N
SMILES: NC1=CC=C(I)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results