accessibility menu, dialog, popup

UserName

CAS RN 135306-45-5

CH3OFF

CAS RN 135306-45-5

IUPAC Name: 1-(3,5-difluorophenyl)propan-1-one
Synonyms: 1-(3,5-difluorophenyl)propan-1-one
Molecular Weight (g/mol): 170.16
Molecular Formula: C9H8F2O
InChi Key: FVDQWXARVQADKN-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC(F)=CC(F)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results