accessibility menu, dialog, popup

UserName

CAS RN 13569-57-8

H2OH2OH2OClIrClCl

CAS RN 13569-57-8

IUPAC Name: trichloroiridium trihydrate
Synonyms: trichloroiridium trihydrate
Molecular Weight (g/mol): 352.61
Molecular Formula: Cl3H6IrO3
InChi Key: LNJXVUXPFZKMNF-UHFFFAOYSA-K
SMILES: O.O.O.Cl[Ir](Cl)Cl
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results