CAS RN 13586-38-4
CAS RN 13586-38-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Ammonium cobalt(II) sulfate hexahydrate, 99%
CAS: 13586-38-4 Molecular Formula: H8CoN2O8S2·6H2O MDL Number: MFCD00151049 Synonym: Cobalt(II) ammonium sulfate hexhydrate; Cobaltous ammonium sulfate hexahydrate
2
Ammonium cobalt(II) sulfate hexahydrate, 98%
CAS: 13586-38-4 Molecular Formula: (NH4)2Co(SO4)2·6H2O MDL Number: MFCD00151049 Synonym: Cobalt(II) ammonium sulfate; Cobaltous ammonium sulfate