missing translation for 'onlineSavingsMsg'
Learn More

Ammonium cobalt(II) sulfate hexahydrate, 99%

Product Code. 11394938
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394938 500 g 500g
11304948 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394938 Supplier Thermo Scientific Alfa Aesar Supplier No. 043902.36
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

As spectroscopic standard, cobalt plating, catalysis, ceramics | CAS: 13586-38-4 | H8CoN2O8S2·6H2O

Ammonium cobalt(II) sulfate hexahydrate is uses as spectroscopic standard, cobalt plating, catalysis and ceramics. It is also used in pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13586-38-4
Molecular Formula H8CoN2O8S2·6H2O
MDL Number MFCD00151049
Synonym Cobalt(II) ammonium sulfate hexhydrate; Cobaltous ammonium sulfate hexahydrate

Specifications

Melting Point ∼120°C (decomposition)
Color Pink to Red
Physical Form Crystalline
Quantity 500 g
Linear Formula (NH4)2Co(SO4)2·6H2O
UN Number UN3077
Solubility Information Soluble in water, Insoluble in alcohol
Formula Weight 395.22 (287.14 Anhydrous)
Percent Purity ≥99%
Odor Odorless
Sensitivity Hygroscopic
Density 1.902 g/mL
Chemical Name or Material Ammonium cobalt (II) sulfate hexahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.