accessibility menu, dialog, popup

UserName

CH3OOCH3Cl

CAS RN 135965-21-8

IUPAC Name: 2-chloro-9,10-bis(4-ethoxyphenyl)anthracene
Synonyms: 2-chloro-9,10-bis(4-ethoxyphenyl)anthracene
Molecular Weight (g/mol): 452.98
Molecular Formula: C30H25ClO2
InChi Key: RFVZBUUPBPFZMH-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=C(OCC)C=C2)C2=CC(Cl)=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results