missing translation for 'onlineSavingsMsg'
Learn More

9,10-Bis(4-ethoxyphenyl)-2-chloroanthracene, 98%, Thermo Scientific™

Product Code. 15420118
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420118 25 g 25g
15400118 1 g 1g
15410118 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15420118 Supplier Thermo Scientific Alfa Aesar Supplier No. H53369.14
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135965-21-8
Molecular Formula C30H25ClO2
Molecular Weight (g/mol) 452.98
MDL Number MFCD00190198
InChI Key RFVZBUUPBPFZMH-UHFFFAOYSA-N
Synonym 2-chloro-9,10-bis 4-ethoxyphenyl anthracene, 9,10-bis 4-ethoxyphenyl-2-chloroanthracene, 9,10-bis-4-ethoxyphenyl-2-chloroanthracene
PubChem CID 21715448
IUPAC Name 2-chloro-9,10-bis(4-ethoxyphenyl)anthracene
SMILES CCOC1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=C(OCC)C=C2)C2=CC(Cl)=CC=C12

Specifications

Melting Point 264°C to 265°C
Quantity 25 g
Formula Weight 452.98
Percent Purity 98%
Chemical Name or Material 9,10-Bis(4-ethoxyphenyl)-2-chloroanthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.