accessibility menu, dialog, popup

UserName

CH3FFF

CAS RN 13630-19-8

IUPAC Name: 1-methyl-2-(trifluoromethyl)benzene
Synonyms: 1-methyl-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 160.14
Molecular Formula: C8H7F3
InChi Key: DVFVNJHIVAPTMS-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results