accessibility menu, dialog, popup

UserName

NH2SOOFF

CAS RN 13656-60-5

IUPAC Name: 2,4-difluorobenzene-1-sulfonamide
Synonyms: 2,4-difluorobenzenesulfonamide
Molecular Weight (g/mol): 193.17
Molecular Formula: C6H5F2NO2S
InChi Key: PTHLPYKPCQLAOF-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(F)C=C1F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results