accessibility menu, dialog, popup

UserName

OOHOBr

CAS RN 136645-25-5

IUPAC Name: 2-[3-(bromomethyl)phenoxy]acetic acid
Synonyms: 3-(bromomethyl)phenoxyacetic acid
Molecular Weight (g/mol): 245.07
Molecular Formula: C9H9BrO3
InChi Key: MSTODKGDFXWAIO-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CC=CC(CBr)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results