accessibility menu, dialog, popup

UserName

SOO

CAS RN 13678-67-6

IUPAC Name: 2-({[(furan-2-yl)methyl]sulfanyl}methyl)furan
Synonyms: 2-{[(furan-2-ylmethyl)sulfanyl]methyl}furan
Molecular Weight (g/mol): 194.25
Molecular Formula: C10H10O2S
InChi Key: UYLKDZXJEKFFHJ-UHFFFAOYSA-N
SMILES: C(SCC1=CC=CO1)C1=CC=CO1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results