missing translation for 'onlineSavingsMsg'
Learn More

Difurfurylsulfide, 96%, Thermo Scientific™

Product Code. 10752291 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Product Code. Quantity Packaging unitSize
10752291 5 g Glass bottle 5g
11916171 25 g Glass bottle 25g
2 options
This item is not returnable. View return policy

Product Code. 10752291

Brand: Thermo Scientific Acros 265080050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 13678-67-6
Molecular Formula C10H10SO2
Molecular Weight (g/mol) 194.25
InChI Key UYLKDZXJEKFFHJ-UHFFFAOYSA-N
Synonym difurfuryl sulfide, furfuryl sulfide, difurfurylsulfide, 2-furfuryl monosulfide, furan, 2,2'-thiobis methylene bis, bis 2-furfuryl sulfide, unii-x6o686cpco, bis furan-2-ylmethyl sulfane, bis-furfuryl sulfide, 2-difurfuryl sulfide
PubChem CID 61659
IUPAC Name 2-(furan-2-ylmethylsulfanylmethyl)furan
SMILES C1=COC(=C1)CSCC2=CC=CO2

Specifications

Melting Point 30.0°C to 32.0°C
Color White to Brown
Boiling Point 135.0°C to 143.0°C (14.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5540 to 1.5560
Quantity 5 g
Beilstein 17,III,355
Solubility Information Solubility in water: insoluble
Formula Weight 194.25
Percent Purity 96%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material Difurfurylsulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.