accessibility menu, dialog, popup

UserName

CAS RN 13709-36-9

FXeF

CAS RN 13709-36-9

IUPAC Name: difluoroxenon
Synonyms: xenon difluoride
Molecular Weight (g/mol): 169.29
Molecular Formula: F2Xe
InChi Key: IGELFKKMDLGCJO-UHFFFAOYSA-N
SMILES: F[Xe]F
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)