missing translation for 'onlineSavingsMsg'
Learn More

Xenon difluoride, 99.5+%

Product Code. 10463215 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Plastic bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10463215 5 g Plastic bottle 5g
10053500 1 g Plastic bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10463215

Brand: Thermo Scientific Acros 278550050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13709-36-9 | F2Xe | 169.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-36-9
Molecular Formula F2Xe
Molecular Weight (g/mol) 169.29
MDL Number MFCD00040538
InChI Key IGELFKKMDLGCJO-UHFFFAOYSA-N
Synonym xenon difluoride, xenon fluoride, xenon fluoride xef2, unii-6poj14981p, bis fluoranyl xenon, xef2, acmc-20aj9j, xenon difluoride trace metals basis
PubChem CID 83674
IUPAC Name difluoroxenon
SMILES F[Xe]F

Specifications

Melting Point 129°C
Color White
Physical Form Crystals
Quantity 5 g
Assay Percent Range 99.5+%
Linear Formula XeF2
Merck Index 15, 10270
Solubility Information Solubility in water: decomposes in water
Formula Weight 169.29
Percent Purity ≥99.5%
Packaging Plastic bottle
Chemical Name or Material Xenon difluoride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Xenon difluoride
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H271-May cause fire or explosion; strong oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.