accessibility menu, dialog, popup

UserName

ClSO

CAS RN 13735-12-1

IUPAC Name: 6-chloro-3,4-dihydro-2H-1-benzothiopyran-4-one
Synonyms: 6-chloro-2,3-dihydro-1-benzothiopyran-4-one
Molecular Weight (g/mol): 198.66
Molecular Formula: C9H7ClOS
InChi Key: OKHUUKHZUNKSQA-UHFFFAOYSA-N
SMILES: ClC1=CC=C2SCCC(=O)C2=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results