missing translation for 'onlineSavingsMsg'
Learn More

6-chloro-3,4-dihydro-2H-1-benzothiin-4-one, 97%, Thermo Scientific™

Product Code. 10314302
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10314302 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10314302 Supplier Thermo Scientific Maybridge Supplier No. SEW00590EA
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 13735-12-1
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C9H7ClOS
MDL Number MFCD00052997
Quantity 10 g
Synonym 6-chlorothiochroman-4-one, 6-chloro-2,3-dihydro-4h-thiochromen-4-one, 6-chloro-3,4-dihydro-2h-1-benzothiopyran-4-one, 6-chloro-thiochroman-4-one, 6-chloro-3,4-dihydro-2h-1-benzothiin-4-one, 6-chloro-2,3-dihydro-4h-1-benzothiopyran-4-one, 6-chloro-2,3-dihydro-1-benzothiopyran-4-one, 6-chloro-2h,3h-benzo e thiin-4-one, acmc-20alz1, maybridge1_007431
InChI Key OKHUUKHZUNKSQA-UHFFFAOYSA-N
SMILES C1CSC2=C(C1=O)C=C(C=C2)Cl
IUPAC Name 6-chloro-2,3-dihydrothiochromen-4-one
Molecular Weight (g/mol) 198.664
PubChem CID 248065
Formula Weight 198.67
Percent Purity 97%
Chemical Name or Material 6-chloro-3, 4-dihydro-2H-1-benzothiin-4-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.