accessibility menu, dialog, popup

UserName

SFFFFFOHCl

CAS RN 1373920-80-9

IUPAC Name: [2-chloro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]methanol
Synonyms: [2-chloro-5-(pentafluoro-λ⁶-sulfanyl)phenyl]methanol
Molecular Weight (g/mol): 268.63
Molecular Formula: C7H6ClF5OS
InChi Key: YRZHFWSMRASOIL-UHFFFAOYSA-N
SMILES: OCC1=C(Cl)C=CC(=C1)S(F)(F)(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results