CAS RN 1375466-18-4
CAS RN 1375466-18-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
ACY-775, MedChemExpress
MedChemExpress ACY-775 is a potent and selective inhibitor of the of histone deacetylase 6 (HDAC6) with an IC50 of 7.5 nM. ACY775 also inhibits metallo-β-lactamase domain-containing protein 2 (MBLAC2).