accessibility menu, dialog, popup

UserName

CAS RN 138716-37-7

IUPAC Name: 5-(4-fluorophenyl)-1,2-oxazole
Synonyms: 5-(4-fluorophenyl)-1,2-oxazole
Molecular Weight (g/mol): 163.15
Molecular Formula: C9H6FNO
InChi Key: JZCBQTRPILCIBC-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C1=CC=NO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results