accessibility menu, dialog, popup

UserName

H3CNCH3H3CH3CSnFF

CAS RN 139353-88-1

IUPAC Name: difluorotriphenylstannanuide; tetrabutylazanium
Synonyms: difluorotriphenylstannanuide; tetrabutylammonium ion
Molecular Weight (g/mol): 630.5
Molecular Formula: C34H51F2NSn
InChi Key: ODMXVCNGZGLSRS-UHFFFAOYSA-L
SMILES: CCCC[N+](CCCC)(CCCC)CCCC.F[Sn-](F)(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results