missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium difluorotriphenyl stannate, 95%, Thermo Scientific™

Product Code. 10502471 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10502471 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10502471 Supplier Thermo Scientific Acros Supplier No. 302590010
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

C-X bond formation reagent, Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

CAS 139353-88-1
Melting Point 187°C to 193°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C34H51F2NSn
MDL Number MFCD00192465
Quantity 1 g
Synonym tetrabutylammonium difluorotriphenylstannate, tetrabutylammonium difluorotriphenylstannate iv, tetrabutylammonium difluorotriphenyltin, tetrabutylammonium triphenyldifluorotin, tetrabutylammoniumdifluorotriphenylstannate, tetrabutylam-monium triphenyldifluorostannate, tetrabutylammonium difluorotriphenyl stannate, tetra-n-butylammonium difluorotriphenylstannate, difluorotriphenylstannanuide; tetrabutylammonium, difluoro triphenyl stannanuide; tetrabutylammonium
InChI Key ODMXVCNGZGLSRS-UHFFFAOYSA-L
SMILES CCCC[N+](CCCC)(CCCC)CCCC.C1=CC=C(C=C1)[Sn-](C2=CC=CC=C2)(C3=CC=CC=C3)(F)F
IUPAC Name difluoro(triphenyl)stannanuide;tetrabutylazanium
Molecular Weight (g/mol) 630.48
PubChem CID 2733194
Formula Weight 630.48
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium difluorotriphenyl stannate, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.