accessibility menu, dialog, popup

UserName

CAS RN 140-11-4

H3COO

CAS RN 140-11-4

IUPAC Name: benzyl acetate
Synonyms: benzyl acetate benzylester kyseliny octove (14C)benzyl acetate benzyl acetate preparation caswell no. 081EA benzyl acetate (natural)
Molecular Weight (g/mol): 150.18
Molecular Formula: C9H10O2
InChi Key: QUKGYYKBILRGFE-UHFFFAOYSA-N
SMILES: CC(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)