missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Acetate, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Benzyl acetate |
| CAS | 140-11-4 |
| Molecular Formula | C9 H10 O2 |
| Quantity | 1 g |
| Synonym | Acetic acid, benzyl ester (6CI,8CI), Benzyl alcohol, acetate (6CI), (Acetoxymethyl)benzene, Benzyl acetate, Benzyl ethanoate, NSC 4550, Phenylmethyl acetate, α-Acetoxytoluene |
| InChI Formula | InChI=1S/C9H10O2/c1-8(10)11-7-9-5-3-2-4-6-9/h2-6H,7H2,1H3 |
| SMILES | CC(=O)OCc1ccccc1 |
| IUPAC Name | benzyl acetate |
| Molecular Weight (g/mol) | 150.17 |
| Formula Weight | 150.0681 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction