accessibility menu, dialog, popup

UserName

CAS RN 14024-61-4

OOPdOOH3CH3CCH3CH3

CAS RN 14024-61-4

IUPAC Name: bis(4-hydroxypent-3-en-2-one) palladium
Synonyms: bis(4-hydroxypent-3-en-2-one) palladium
Molecular Weight (g/mol): 306.65
Molecular Formula: C10H16O4Pd
InChi Key: BABLLCDZHABSRT-UHFFFAOYSA-N
SMILES: [Pd].CC(O)=CC(C)=O.CC(O)=CC(C)=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (4)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)