missing translation for 'onlineSavingsMsg'
Learn More

Palladium(II) Acetylacetonate, TRC

Product Code. 30418215
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Product Code. Quantity unitSize
30418215 250 mg 250mg
1 options
This item is not returnable. View return policy

Product Code. 30418215

Brand: Toronto Research Chemicals TRCP578015250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Palladium(II) Acetylacetonate
CAS 14024-61-4
Molecular Formula 2 C5 H7 O2 . Pd
Quantity 250 mg
Synonym (SP-4-1)-Bis(2,4-pentanedionato-κO2,κO4)palladium, Bis(2,4-pentanedionato)palladium, Bis(2,4-pentanedionato-κO,κO')palladium, Bis(acetylacetonato)palladium, Bis(acetylacetonato)palladium(II), NSC 187660, Palladium Acetylacetonate, Palladium bis(2,4-pentanedionate), Palladium Bis(acetoacetonate), Palladium Bis(acetylacetonate), Palladium(II) Acetylacetonate, Pd (II) Diacetylacetonate, Bis(2,4-pentanedionato-κO,κO')-','(SP-4-1)-Palladium','Bis(2,4-pentanedionato)-palladium
InChI Formula InChI=1S/2C5H7O2.Pd/c2*1-4(6)3-5(2)7;/h2*3H,1-2H3;/q2*-1;+2
SMILES [Pd+2].CC(=O)[CH-]C(=O)C.CC(=O)[CH-]C(=O)C
IUPAC Name palladium(2+);pentane-2,4-dione
Molecular Weight (g/mol) 304.636
Formula Weight 303.993
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.