accessibility menu, dialog, popup

UserName

H3COOBr

CAS RN 14062-25-0

IUPAC Name: ethyl 2-(4-bromophenyl)acetate
Synonyms: ethyl 2-(4-bromophenyl)acetate
Molecular Weight (g/mol): 243.1
Molecular Formula: C10H11BrO2
InChi Key: ZFDCWHPNBWPPHG-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CC=C(Br)C=C1
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)