accessibility menu, dialog, popup

UserName

CH3SCH3O

CAS RN 14109-72-9

IUPAC Name: 1-(methylsulfanyl)propan-2-one
Synonyms: 1-(methylsulfanyl)propan-2-one
Molecular Weight (g/mol): 104.17
Molecular Formula: C4H8OS
InChi Key: UKFADLGENFFWHR-UHFFFAOYSA-N
SMILES: CSCC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)