missing translation for 'onlineSavingsMsg'
Learn More

1-Methylthio-2-propanone, 98+%

Product Code. 11441167
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11441167 25 g 25g
11431167 5 g 5g
11424160 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11441167 Supplier Thermo Scientific Alfa Aesar Supplier No. B24149.14
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14109-72-9 | C4H8OS | 104.17 g/mol

1-Methylthio-2-propanone is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylthio-2-propanone is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Soluble in alcohol and slightly soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14109-72-9
Molecular Formula C4H8OS
Molecular Weight (g/mol) 104.17
MDL Number MFCD00015325
InChI Key UKFADLGENFFWHR-UHFFFAOYSA-N
Synonym 1-methylthio-2-propanone, methylthio acetone, 2-thia-4-pentanone, unii-s6v0dw7bqa, 1-methylsulfanyl propan-2-one, 2-propanone, 1-methylthio, methylthio-2-propanone, acetonyl methyl sulfide, 1-methylsulfanyl acetone, s6v0dw7bqa
PubChem CID 6430713
IUPAC Name 1-methylsulfanylpropan-2-one
SMILES CSCC(C)=O

Specifications

Density 1.023
Boiling Point 69°C to 70°C (30 mmHg)
Flash Point 43°C (109°F)
Odor Stench
Refractive Index 1.474
Quantity 25 g
UN Number UN1224
Beilstein 1737738
Solubility Information Soluble in alcohol and slightly soluble in water.
Formula Weight 104.17
Percent Purity ≥98%
Chemical Name or Material 1-Methylthio-2-propanone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.