accessibility menu, dialog, popup

UserName

CH3OCl

CAS RN 14123-60-5

IUPAC Name: 1-(3-chlorophenyl)propan-2-one
Synonyms: 1-(3-chlorophenyl)propan-2-one
Molecular Weight (g/mol): 168.62
Molecular Formula: C9H9ClO
InChi Key: VCNYPJMEQHTAHS-UHFFFAOYSA-N
SMILES: CC(=O)CC1=CC=CC(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results