CAS RN 1418095-19-8
CAS RN 1418095-19-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(2-Ethyl-6-methylphenyl)-N-(2-sulfoacetyl)-L-alanine, TRC
CAS: 1418095-19-8 Molecular Formula: C14 H19 N O6 S Molecular Weight (g/mol): 329.37 Synonym: (S)-2-(N-(2-Ethyl-6-methylphenyl)-2-sulfoacetamido)propanoic Acid,N-(2-Ethyl-6-methylphenyl)-N-(2-sulfoacetyl)-L-alanine IUPAC Name: (2S)-2-(2-ethyl-6-methyl-N-(2-sulfoacetyl)anilino)propanoic acid SMILES: CCc1cccc(C)c1N([C@@H](C)C(=O)O)C(=O)CS(=O)(=O)O