accessibility menu, dialog, popup

UserName

NaH3COSOOO

CAS RN 142-87-0

IUPAC Name: sodium decyl sulfate
Synonyms: sodium decyl sulfate
Molecular Weight (g/mol): 260.32
Molecular Formula: C10H21NaO4S
InChi Key: XZTJQQLJJCXOLP-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCCOS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (2)
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (4)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)