missing translation for 'onlineSavingsMsg'
Learn More

Sodium Decyl Sulfate, TRC

Product Code. 30136845
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136845 10 g 10g
30134162 5 g 5g
30009077 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136845 Supplier Toronto Research Chemicals Supplier No. TRCS22459510G
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-87-0
Molecular Formula C10H21O4SNa
Molecular Weight (g/mol) 260.33
Synonym Avirol 110, Compound 132D, DSS, DSS (Surfactant), Decyl Sulfate Sodium Salt;Emal 3F, Pentrone DN, Polystep B 25, SDeS, Sodium Capryl Sulfate, Sodium Decyl Sulfate, Sodium Hydrogen Sulfate Decyl Ester, Sodium n-Decyl Sulfate, Sulfochem NADS, Sulfotex 110;
IUPAC Name sodium;decyl sulfate
SMILES [Na+].CCCCCCCCCCOS(=O)(=O)[O-]

Specifications

Quantity 10 g
InChI Formula InChI=1S/C10H22O4S.Na/c1-2-3-4-5-6-7-8-9-10-14-15(11,12)13;/h2-10H2,1H3,(H,11,12,13);/q;+1/p-1
Formula Weight 260.1058
Chemical Name or Material Sodium Decyl Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.