accessibility menu, dialog, popup

UserName

H3COOCH3CH2

CAS RN 142-90-5

IUPAC Name: dodecyl 2-methylprop-2-enoate
Synonyms: dodecyl methacrylate
Molecular Weight (g/mol): 254.41
Molecular Formula: C16H30O2
InChi Key: GMSCBRSQMRDRCD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOC(=O)C(C)=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)