missing translation for 'onlineSavingsMsg'
Learn More

Lauryl methacrylate, 97%, stabilized

Product Code. 10294891 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10294891 100 mL Glass bottle 100mL
10692942 500 mL Glass bottle 500mL
11924961 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10294891 Supplier Thermo Scientific Acros Supplier No. 347711000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 142-90-5 | C16H30O2 | 254.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-90-5
Molecular Formula C16H30O2
Molecular Weight (g/mol) 254.41
MDL Number MFCD00008972
InChI Key GMSCBRSQMRDRCD-UHFFFAOYSA-N
Synonym dodecyl methacrylate, lauryl methacrylate, dodecyl 2-methylacrylate, metazene, n-dodecyl methacrylate, sipomer lma, 2-propenoic acid, 2-methyl-, dodecyl ester, dodecyl 2-methyl-2-propenoate, lama, ageflex fm 246
PubChem CID 8906
IUPAC Name dodecyl 2-methylprop-2-enoate
SMILES CCCCCCCCCCCCOC(=O)C(=C)C

Specifications

Melting Point -23.0°C
Color Yellow
Density 0.8710g/mL
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH2=C(CH3)COO(CH2)11CH3
Refractive Index 1.4440 to 1.4470
Quantity 100 mL
Specific Gravity 0.871
Solubility Information Solubility in water: insoluble
Stabilizer 300 to 500ppm MEHQ
Viscosity 4.7 mPa.s (25°C)
Formula Weight 254.41
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Lauryl methacrylate, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.