accessibility menu, dialog, popup

UserName

H2NNN

CAS RN 142335-61-3

IUPAC Name: 4-[(1H-pyrazol-1-yl)methyl]aniline
Synonyms: 4-(pyrazol-1-ylmethyl)aniline
Molecular Weight (g/mol): 173.22
Molecular Formula: C10H11N3
InChi Key: ISRYTHBRUSOVAB-UHFFFAOYSA-N
SMILES: NC1=CC=C(CN2C=CC=N2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results