missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Pyrazol-1-ylmethyl)aniline, 95%, Thermo Scientific™

Product Code. 10476853
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10476853 1 g 1g
10420754 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10476853 Supplier Thermo Scientific Maybridge Supplier No. CC56014DA
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 142335-61-3
Molecular Formula C10H11N3
Molecular Weight (g/mol) 173.22
MDL Number MFCD03422514
InChI Key ISRYTHBRUSOVAB-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-ylmethyl aniline, 4-1h-pyrazol-1-yl methyl aniline, 4-pyrazol-1-ylmethyl aniline, 4-1-pyrazolyl methyl aniline, 4-pyrazol-1-ylmethyl-phenylamine, 4-pyrazol-1-ylmethylphenylamine, benzenamine,4-1h-pyrazol-1-ylmethyl, 4-pyrazolylmethyl phenylamine, pyrazolylmethylaniline
PubChem CID 2764412
IUPAC Name 4-(pyrazol-1-ylmethyl)aniline
SMILES NC1=CC=C(CN2C=CC=N2)C=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 173.22
Percent Purity 95%
Chemical Name or Material 4-(1H-Pyrazol-1-ylmethyl)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.