accessibility menu, dialog, popup

UserName

CAS RN 142335-64-6

H2NNH

CAS RN 142335-64-6

IUPAC Name: 1-[(4-aminophenyl)methyl]pyrrolidin-1-ium
Synonyms: 1-[(4-aminophenyl)methyl]pyrrolidin-1-ium
Molecular Weight (g/mol): 177.27
Molecular Formula: C11H17N2
InChi Key: SFEAIUCOZWDYMJ-UHFFFAOYSA-O
SMILES: NC1=CC=C(C[NH+]2CCCC2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results