CAS RN 1424-66-4
IUPAC Name:
2-chloro-4-(dimethylamino)benzaldehyde
Synonyms:
2-chloro-4-(dimethylamino)benzaldehyde
Molecular Weight (g/mol):
183.64
Molecular Formula:
C9H10ClNO
InChi Key:
XSQFAWMDRFSIMY-UHFFFAOYSA-N
SMILES:
CN(C)C1=CC=C(C=O)C(Cl)=C1
1
–
1
of
1
results
1
2-Chloro-4-(dimethylamino)benzaldehyde, ≥95%, Thermo Scientific™
CAS: 1424-66-4 Molecular Formula: C9H10ClNO Molecular Weight (g/mol): 183.64 MDL Number: MFCD00021051 InChI Key: XSQFAWMDRFSIMY-UHFFFAOYSA-N Synonym: 2-chloro-4-dimethylamino benzaldehyde,benzaldehyde, 2-chloro-4-dimethylamino,benzaldehyde,2-chloro-4-dimethylamino,2-chloro-4-n,n-bis methyl amino benzaldehyde,4-dimethylamino-2-chlorobenzaldehyde,acmc-209coh,3-chloro-n,n-dimethyl-4-formylaniline PubChem CID: 74021 IUPAC Name: 2-chloro-4-(dimethylamino)benzaldehyde SMILES: CN(C)C1=CC=C(C=O)C(Cl)=C1