CAS RN 14265-44-2
Buffer Solution, pH 12.00, Chem-Lab
Designed for pH measurement and maintenance in alkaline samples. Buffer Solution, pH 12.00, Chem-Lab is a solution of phosphate and sodium hydroxide, with a pH level of 12.00 ±0.02 at 20°C. Solution is traceable to standard reference materials (SRM) from NIST.
Phosphate as P @ 1000 μg/mL in H2O, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Phosphate Standard: PO4(-3) @ 1000 μg/mL in H2O, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Phosphate as P @ 10000 μg/mL in H2O, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Phosphate Standard: PO4(-3) @ 10000 μg/mL in H2O, VHG™
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications