accessibility menu, dialog, popup

UserName

OHBrBrOOBr

CAS RN 14348-40-4

IUPAC Name: 2,4,6-tribromo-3-hydroxybenzoate
Synonyms: 2,4,6-tribromo-3-hydroxybenzoate
Molecular Weight (g/mol): 373.8
Molecular Formula: C7H2Br3O3
InChi Key: YDBHVMTTYXWHLI-UHFFFAOYSA-M
SMILES: OC1=C(Br)C=C(Br)C(C([O-])=O)=C1Br
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results