accessibility menu, dialog, popup

UserName

O(E)

CAS RN 14371-10-9

IUPAC Name: (2E)-3-phenylprop-2-enal
Synonyms: cinnamal aldehyd skoricovy epa pesticide chemical code 040506 abion CA fema number 2286 supercinnamaldehyde cinnemaldehyde 3-fenylpropenal cinnamic aldehyde, (E)-isomer hefty dog and cat repellent
Molecular Weight (g/mol): 132.16
Molecular Formula: C9H8O
InChi Key: KJPRLNWUNMBNBZ-QPJJXVBHSA-N
SMILES: O=C\C=C\C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (4)
  • (3)
  • (4)
  • (7)