accessibility menu, dialog, popup

UserName

N

CAS RN 14377-68-5

IUPAC Name: 1-phenylcyclobutane-1-carbonitrile
Synonyms: 1-phenylcyclobutane-1-carbonitrile
Molecular Weight (g/mol): 157.22
Molecular Formula: C11H11N
InChi Key: DHIDUDPFTZJPCQ-UHFFFAOYSA-N
SMILES: N#CC1(CCC1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results