missing translation for 'onlineSavingsMsg'
Learn More

1-Phenylcyclobutanecarbonitrile, 95%, Thermo Scientific™

Product Code. 10645782 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10645782 25 g Glass bottle 25g
10635782 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10645782 Supplier Thermo Scientific Acros Supplier No. 154950250
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 14377-68-5
MDL Number MFCD00019258
InChI Key DHIDUDPFTZJPCQ-UHFFFAOYSA-N
Synonym 1-phenylcyclobutanecarbonitrile, cyclobutanecarbonitrile, 1-phenyl, 1-phenyl-cyclobutanecarbonitrile, acmc-1c523, cyclobutanecarbonitrile,1-phenyl, 1-phenyl-1-cyclobutanecarbonitrile, 1-phenyl-1-cyclobutane carbonitrile
PubChem CID 84400
IUPAC Name 1-phenylcyclobutane-1-carbonitrile
SMILES C1CC(C1)(C#N)C2=CC=CC=C2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow
Density 1.0300g/mL
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Molecular Formula C11H11N
Linear Formula C6H5C4H6CN
Refractive Index 1.5313 to 1.5333
Quantity 25 g
Specific Gravity 1.03
Molecular Weight (g/mol) 157.21
Formula Weight 157.21
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 1-Phenylcyclobutanecarbonitrile, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 1-Phenylcyclobutanecarbonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.