CAS RN 143860-04-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Ethyl-2-methyl-2-(3-methylbutyl)-oxazolidine, TRC
CAS: 143860-04-2 Molecular Formula: C11 H23 N O Molecular Weight (g/mol): 185.31 Synonym: Oxazolidine, 3-ethyl-2-methyl-2-(3-methylbutyl)-,3-Ethyl-2-methyl-2-(3-methylbutyl)oxazolidine,3-Ethyl-2-methyl-2-(3-methylbutyl)-1,3-oxazolidine,MS-PLUS,Zoldine MS-PLUS IUPAC Name: 3-ethyl-2-methyl-2-(3-methylbutyl)-1,3-oxazolidine SMILES: CCN1CCOC1(C)CCC(C)C