missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-2-methyl-2-(3-methylbutyl)-oxazolidine, TRC

Product Code. 30010535
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010535 1 g 1g
30007807 100 mg 100mg
30004834 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010535 Supplier Toronto Research Chemicals Supplier No. TRCE6796301G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143860-04-2
Molecular Formula C11 H23 N O
Molecular Weight (g/mol) 185.31
Synonym Oxazolidine, 3-ethyl-2-methyl-2-(3-methylbutyl)-, 3-Ethyl-2-methyl-2-(3-methylbutyl)oxazolidine, 3-Ethyl-2-methyl-2-(3-methylbutyl)-1,3-oxazolidine, MS-PLUS, Zoldine MS-PLUS
IUPAC Name 3-ethyl-2-methyl-2-(3-methylbutyl)-1,3-oxazolidine
SMILES CCN1CCOC1(C)CCC(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C11H23NO/c1-5-12-8-9-13-11(12,4)7-6-10(2)3/h10H,5-9H2,1-4H3
Formula Weight 185.178
Percent Purity >95%
Chemical Name or Material 3-Ethyl-2-methyl-2-(3-methylbutyl)-oxazolidine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.