accessibility menu, dialog, popup

UserName

HClH2NNHCl

CAS RN 14401-51-5

IUPAC Name: hydrogen 4-chlorobenzene-1-carboximidamide chloride
Synonyms: hydrogen chlorophenamidine chloride
Molecular Weight (g/mol): 191.06
Molecular Formula: C7H8Cl2N2
InChi Key: RXAOGVQDNBYURA-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC(=N)C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)