missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzamidine hydrochloride, 95%, Thermo Scientific™

Product Code. 13595020 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13595020 5 g 5g
13515030 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 13595020 Supplier Thermo Scientific Acros Supplier No. 448660050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 14401-51-5
Molecular Formula C7H7ClN2·ClH
Molecular Weight (g/mol) 191.06
InChI Key RXAOGVQDNBYURA-UHFFFAOYSA-N
Synonym 4-chlorobenzamidine hydrochloride, 4-chlorobenzene-1-carboximidamide hydrochloride, 4-chlorobenzamidine hcl, p-chlorobenzamidine hydrochloride, 4-chloro-benzamidine hydrochloride, 4-chlorobenzenecarboximidamide monohydrochloride, benzenecarboximidamide, 4-chloro-, monohydrochloride, 4-chlorobenzimidamide hydrochloride, benzamide, p-chloro-, hydrochloride 7ci, benzamidine, p-chloro-, monohydrochloride
PubChem CID 203436
IUPAC Name 4-chlorobenzenecarboximidamide;hydrochloride
SMILES C1=CC(=CC=C1C(=N)N)Cl.Cl

Specifications

Melting Point 244.0°C to 245.0°C
Quantity 5 g
Packaging Glass bottle
Assay Percent Range 94 to max. 106% min. (Argentometry)
Linear Formula ClC6H4C(=NH)NH2·HCl
Formula Weight 191.06
Percent Purity 95%
Chemical Name or Material 4-Chlorobenzamidine hydrochloride
compliance-icons
Product Identifier
  • 4-Chlorobenzamidine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.