accessibility menu, dialog, popup

UserName

OHFFBr

CAS RN 144292-32-0

IUPAC Name: 4-bromo-2,3-difluorophenol
Synonyms: 4-bromo-2,3-difluorophenol
Molecular Weight (g/mol): 208.99
Molecular Formula: C6H3BrF2O
InChi Key: JZAVCMMYGSROJP-UHFFFAOYSA-N
SMILES: OC1=C(F)C(F)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results